Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HCCS Rabbit Polyclonal Antibody (HRP)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15424R-HRP
BOSSBS-15424R-HRPEA 900 EUR InStock
BOSSBS-15424R-HRP
Anti-HCCS Rabbit Polyclonal Antibody (HRP)
Antikörper
HCCS is a 79 amino acid protein that may act as a tumor suppressor, promoting the apoptosis of cancer cells. Expressed in leukocytes, lung, spleen, liver, heart, kidney, muscle and uterine cervix, HCCS-1 is down-regulated in cervical cancers. The gene encoding HCCS-1 maps to human chromosome 15q25.1. Encoding more than 700 genes, chromosome 15 is made up of approximately 106 million base pairs and comprises about 3% of the human genome. Angelman and Prader-Willi syndromes are associated with loss of function or deletion of genes in the 15q11-q13 region. Tay-Sachs disease is a lethal disorder associated with mutations of the HEXA gene, which is encoded by chromosome 15. Marfan syndrome is associated with chromosome 15 through the FBN1 gene.

Type: Primary
Antigen: HCCS
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 51-150/268
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge