Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HCP5 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15429R-A350
BOSSBS-15429R-A350EA 893 EUR InStock
BOSSBS-15429R-A350
Anti-HCP5 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
Antikörper
HCP5 is a region present on chromosome 6p21.3 that is characterised by multiple duplicated gene families. HCP5 (HLA class I histocompatibility antigen protein P5), also known as P5-1, is a 132 amino acid protein that is encoded by a gene mapping to human chromosome 6p21.33. Localising within the MHC class I region, HCP5 is not structurally related to other MHC class I genes, but does have high sequence similarity with HERV-L and HERV-16. HCP5 also has high sequence homology to retroviral Pol genes, making it a possible candidate for interaction with HIV-1 through an antisense mechanism that prevents retrovirus transcription. Single-nucleotide polymorphisms (SNPs) to the region of the HCP5 gene that corresponds with HLA-B has been linked to a lower HIV-1 viral set point. HCP5 is highly expressed in lymphoid tissues, spleen and activated lymphocytes, as well as B-cell and natural killer (NK) cell lines.

Type: Primary
Antigen: HCP5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 1-80/132
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge