Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HDPTP Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15434R-A350
BOSSBS-15434R-A350EA 893 EUR InStock
BOSSBS-15434R-A350
Anti-HDPTP Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
Antikörper
HD-PTP is a 1636 amino acid protein encoded by the human gene PTPN23. HD-PTP belongs to the protein-tyrosine phosphatase family, non-receptor class subfamily. It contains one BRO1 domain, two TPR repeats and one tyrosine-protein phosphatase domain. The C-terminal region contains the PTP-like domain, whereas the N-terminal region contains the two TPR regions. These regions are homologous to the yeast protein, BRO1, which is involved in the mitogen-activated protein kinase signaling pathway. Similarly, HD-PTP is believed to act as a negative regulator of Ras-mediated mitogenic activity and is phosphorylated upon DNA damage, probably by ATM or ATR. HD-PTP protein is differentially modulated by two angiogenic growth factors. While Vascular Endothelial Growth Factor (VEGF) has no affect on protein levels, Fibroblast Growth Factor-2 (FGF-2) induces HD-PTP degradation via the proteasome system.

Type: Primary
Antigen: HDPTP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 1501-1636/1636
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge