Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HELZ Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15450R
BOSSBS-15450REA 725 EUR InStock
BOSSBS-15450R
Anti-HELZ Rabbit Polyclonal Antibody
Antikörper
Helicases comprise a class of enzymes that function as motor proteins which move along nucleic acid phosphodiester bonds, effectively separating two annealed nucleic acid strands. RNA helicases alter the conformation of RNA, specifically by unwinding double-stranded RNA regions to yield single RNA strands, a process which changes the biological activity of the RNA molecule. HELZ (helicase with zinc finger), also known as DHRC or HUMORF5, is a 1,942 amino acid nuclear protein that contains one C3H1-type zinc finger and belongs to the RNA helicase superfamily. Expressed ubiquitously during embryonic development, HELZ is thought to function as an RNA helicase that modifies RNA structure and plays a role in the development of multiple organs and tissues within the developing embryo.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HELZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge