Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HBcAg Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15455R-HRP
BOSSBS-15455R-HRPEA 1350 EUR InStock
BOSSBS-15455R-HRP
Anti-HBcAg Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Hepatitis B Virus Core Antigen (HBcAg) is part of the infectious virion containing an inner 'core particle' enclosing the viral genome. The icosahedral core particle contains 180 or 240 copies of the core protein. HBcAg is one of the three major clinical antigens of hepatitis B virus but disappears early in the course of infection.The hepatitis B virus core antigen (HBcAg) is a highly immunogenic subviral particle and functions as both a T-cell-dependent and a T-cell-independent antigen. Therefore, HBcAg may be a promising candidate target for therapeutic vaccine control of chronic HBV infection.

Type: Primary
Antigen: HBcAg
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 1-100/185
Host: Rabbit
Isotype: IgG
Reactivity: Virus

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge