Anti-HGD Rabbit Polyclonal Antibody (FITC)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-15472R-FITC
BOSSBS-15472R-FITCEA 900 EUR InStock
BOSSBS-15472R-FITC
Anti-HGD Rabbit Polyclonal Antibody (FITC)
Antikörper
HGD is a 445 amino acid protein that belongs to the homogentisate dioxygenase family and is involved in the pathway of amino acid degradation. Expressed at high levels in kidney, colon, liver, prostate and small intestine, HGD uses iron as a cofactor to catalyse the oxygen-dependent conversion of homogentisate to 4-maleylacetoacetate, a reaction that is the fourth step in the creation of L-phenylalanine from fumarate and acetoacetic acid. Defects in the gene encoding HGD are the cause of alkaptonuria (AKU), an autosomal recessive disorder that is characterised by urine that turns dark on standing and alkalinisation, black ochronotic pigmentation of cartilage and collagenous tissues and spine arthritis.
Type: Primary
Antigen: HGD
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 351-445/445
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge