Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HHAT Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15474R-A750
BOSSBS-15474R-A750EA 890 EUR InStock
BOSSBS-15474R-A750
Anti-HHAT Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antikörper
HHAT is a 493 amino acid multi-pass membrane protein that localises to the endoplasmic reticulum and belongs to the membrane-bound acyltransferase family. Expressed ubiquitously, HHAT functions to catalyse the N-terminal palmitoylation of SSH (slingshot homolog), an event that is required for SHH signaling pathways. HHAT is expressed in cancer cell lines, suggesting a role for HHAT in tumorigenesis. The gene encoding HHAT maps to human chromosome 1 and is expressed as four alternatively spliced isoforms. Chromosome 1 is the largest human chromosome, spanning about 260 million base pairs and making up 8% of the human genome. Several disorders, including Stickler syndrome, Parkinsons Disease, Gaucher disease, malignant melanoma and Usher syndrome, are caused by defects in genes that localise to chromosome 1.

Type: Primary
Antigen: HHAT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/493
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge