Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HHCM Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15475R
BOSSBS-15475REA 725 EUR InStock
BOSSBS-15475R
Anti-HHCM Rabbit Polyclonal Antibody
Antikörper
HHCM is a 467 amino acid protein that has oncogenic transforming capabilities on a variety of different cell lines. When HHCM was transfected into BRL-1 (Buffalo rat liver) or NIH3T3 cells, the cells exhibited significant morphological changes, anchorage independent growth and loss of contact inhibition. When the cells were inoculated into rats and mice, they became highly tumorigenic. HHCM gene-related DNA sequences were found in at least nineteen hepatomas of Asian origin. The gene encoding HHCM maps to human chromosome 8, which is made up of nearly 146 million bases and encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and are typically associated with a poor prognosis.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HHCM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge