Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HIBADH Rabbit Polyclonal Antibody (FITC)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15483R-FITC
BOSSBS-15483R-FITCEA 900 EUR InStock
BOSSBS-15483R-FITC
Anti-HIBADH Rabbit Polyclonal Antibody (FITC)
Antikörper
HIBADH is a 336 amino acid mitochondrial enzyme that catalyses the NAD+-dependent, reversible oxidisation of 3-Hydroxyisobutyrate to methylmalonate semialdehyde, an intermediate of valine catabolism. The enzyme functions as a homodimer between a pH of 7,0 and 10,0, with optimal activity between 8,8 and 9,0. It was previously hypothesised that defects in the gene encoding HIBADH may be the cause of 3-Hydroxyisobutyric aciduria, a rare disorder that is characterised by a variety of clinical manifestations such as neurodevelopmental problems and dysmorphic features. However, it was shown that HIBADH activity was equal in patients with 3-Hydroxyisobutyric aciduria as compared with controls.

Type: Primary
Antigen: HIBADH
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 251-336/336
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge