Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HIBCH Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15484R-A750
BOSSBS-15484R-A750EA 890 EUR InStock
BOSSBS-15484R-A750
Anti-HIBCH Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antikörper
HIBCH is a 386 amino acid protein belonging to the enoyl-CoA hydratase/isomerase family. Localising to the mitochondria, HIBCH is highly expressed in liver and kidney, with lower levels found in heart, muscle and brain. HIBCH hydrolyses HIBYL-CoA, a saline catabolite, and _-hydroxypropionyl-CoA, an intermediate in the minor pathway involved in the metabolism of proprionate. Existing as two alternatively spliced isoforms, the gene encoding HIBCH maps to human chromosome 2q32.2. Defects to this gene result in HIBCH deficiency (HIBCHD), known alternatively as deficiency of _-hydroxyisobutyryl CoA deacylase or methacrylic aciduria. HIBCHD is characterised by the accumulation of methacrylyl-CoA, a highly reactive compound that undergoes addition reactions with free sulfhydryl groups. Phenotypic symptoms include early deterioration of neurological function, delayed motor skill development and hypotonia.

Type: Primary
Antigen: HIBCH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/386
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge