Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HIC2 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15486R
BOSSBS-15486REA 725 EUR InStock
BOSSBS-15486R
Anti-HIC2 Rabbit Polyclonal Antibody
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krüppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. HIC-2 (hypermethylated in cancer 2) possesses zinc finger motifs that are thought to be important for DNA-binding and also has a BTB/POZ domain at the N-terminus, which is thought to be important for protein-protein binding, as well as for the binding of transcription factors. HIC-2 is also known as Hic-3, HIC1-related gene on chromosome 22 or Zinc finger and BTB domain-containing protein 30, and is a 615 amino acid protein that is expressed as two isoforms produced by alternative splicing. HIC-2 is highly expressed in cerebellum and is localized to the nucleus in cells. HIC-2 contains a short amino acid sequence that is thought to interact with CtBP, a transcriptional repressor. The gene sequence associated with HIC-2 is thought to be a target for miRNAs (microRNAs) which are expressed in many cancers, suggesting that HIC-2 could possess tumor suppressor capabilities.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HIC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge