Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PCSK4 Rabbit Polyclonal Antibody (Cy5)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15501R-CY5
BOSSBS-15501R-CY5EA 900 EUR InStock
BOSSBS-15501R-CY5
Anti-PCSK4 Rabbit Polyclonal Antibody (Cy5)
Antikörper
PCSK4 is a member of the subtilisin-like proprotein convertase family. These enzymes process latent precursor proteins into their biologically active products. The encoded protein plays a critical role in reproduction and processes multiple prohormones including pro-pituitary adenylate cyclase-activating protein (proPACAP) and pro-insulin-like growth factor II.

Type: Primary
Antigen: PCSK4
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 221-320/755
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge