Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ITM5 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15518R-A488
BOSSBS-15518R-A488EA 893 EUR InStock
BOSSBS-15518R-A488
Anti-ITM5 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Antikörper
IFITM5 is a membrane protein thought to play a role in bone mineralisation. This gene is located on chromosome 11 in a cluster of related genes which are induced by interferon, however, this gene has not been shown to be interferon inducible. A similar gene, located in a gene cluster on mouse chromosome 7, is a member of the interferon-inducible fragilis gene family. The mouse gene encodes a transmembrane protein described as participating in germ cell competence. A mutation in the 5' UTR of this gene has been associated with osteogenesis imperfecta type V (PMID: 22863190, 22863195).

Type: Primary
Antigen: ITM5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 1-100/132
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge