Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-alpha Adducin Ser436 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-15968R-A350
BOSSBS-15968R-A350EA 921 EUR InStock
BOSSBS-15968R-A350
Anti-alpha Adducin Ser436 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
Antikörper
Adducins are a family of cytoskeleton proteins encoded by three genes (alpha, beta, gamma). Adducin is a protein associated with the inner leaflet of the plasma membrane and is one of the proteins localised at the spectrin-actin junction of the membrane skeleton. The cortical actin cytoskeletal network is lost during apoptosis and Adducins are central in the cortical actin network organisation. Adducin Alpha is a cytoskeletal protein involved with sodium-pump activity in the renal tubule and is associated with hypertension. The expression of Adducin Alpha and Adducin Gamma is ubiquitous in contrast to the restricted expression of Adducin Beta. Adducin Beta is expressed at high levels in brain and hematopoietic tissues, such as bone marrow, in humans, and in spleen in mice.

Type: Primary
Antigen: alpha Adducin Ser436
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge