Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Parafibromin Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1597R-A555
BOSSBS-1597R-A555EA 893 EUR InStock
BOSSBS-1597R-A555
Anti-Parafibromin Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The HRPT2 gene product, parafibromin, is a tumor suppressor protein that is part of the human Paf1 complex. The yeast counterpart to this complex is part of the RNA polymerase II complex as well, and is important for histone modification and connections to posttranscriptional events. Human parafibromin also associates with the RNA polymerase II large subunit.

Type: Primary
Antigen: Parafibromin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 61-160/531
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge