Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FOX3/NeuN Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1613R-CY3
BOSSBS-1613R-CY3EA 909 EUR InStock
BOSSBS-1613R-CY3
Anti-FOX3/NeuN Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Vertebrate neuron-specific nuclear protein called NeuN (Neuronal Nuclei) is an excellent marker for neurons in primary cultures and in retinoic acid-stimulated P19 cells. It is also useful for identifying neurons in transplants. NeuN is a neuron-specific, DNA-binding nuclear protein in vertebrates. In mice, NeuN is observed in most neuronal cell types throughout the nervous system, including cerebellum, cerebral cortex, hippocampus, thalamus and spinal cord, as well as the dorsal root ganglia, sympathetic chain ganglia and enteric ganglia of the peripheral nervous system. NeuN immunoreactivity is first observed in neurons when they become post-mitotic and are initiating cellular and morphological differentiation. No staining is observed in proliferative zones. NeuN has been used as an immunohistochemical marker for excitotoxic lesions of the brain as well as in the diagnosis of a wide range of human tissue specimens from the central and peripheral nervous systems.

Type: Primary
Antigen: FOX3/NeuN
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 51-150/312
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge