Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DEFA1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1697R-CY5.5
BOSSBS-1697R-CY5.5EA 900 EUR InStock
BOSSBS-1697R-CY5.5
Anti-DEFA1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Defensins are a group of cyclic peptides containing 29-35 amino acids, which comprise approximately 30% of the total protein content of neutrophil azurophilic granules. Various functions have been reported for the defensins, including antibacterial and antifungal activity and chemotactic activity for monocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DEFA1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge