Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HPV16 E6 + HPV18 E6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1719R-HRP
BOSSBS-1719R-HRPEA 900 EUR InStock
BOSSBS-1719R-HRP
Anti-HPV16 E6 + HPV18 E6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Transcriptional transactivator. Binds double-stranded DNA (By similarity). Has transforming activity. Inactivates, with E6-AP ubiquitin-protein ligase, the human p53/TP53 tumor suppressor protein by targeting it to degradation. Binds and targets human MUPP1/MPDZ protein to degradation. Those two functions presumably contribute to transforming activity. Interaction with human FBLN1 protein also seems to be linked to cell transformation.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: HPV16 E6 + HPV18 E6
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge