Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LOC494224 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1766R-A647
BOSSBS-1766R-A647EA 893 EUR InStock
BOSSBS-1766R-A647
Anti-LOC494224 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Follicular dendritic cells are cells that facilitate antigen recognition by B cells in follicles. This kind of dendritic cell is not bone marrow derived and is not a kind of blood cell. It is purely a resident of the follicles of secondary lymphoid organs. B cells form germinal centers around follicular dendritic cells in lymphoid organs. Dendritic cells form from monocytes, white blood cells which circulate in the body and, depending on the right signal, can turn into dendritic cells or macrophages. The monocytes in turn are formed from stem cells in the bone marrow.In normal tissue this antibody identifies dendritic cells and a proportion of B lymphocytes. Evidence in pathological tissue and functional studies suggests it binds to an epitope expressed by antigen presenting cells. Within the macrophage/dendritic cell populations the epitope seen by RFD1 is coexpressed with RFD7 by a subset of cells that exhibit suppressive activity on T cell stimulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LOC494224
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge