Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HMGB4 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1820R-CY3
BOSSBS-1820R-CY3EA 900 EUR InStock
BOSSBS-1820R-CY3
Anti-HMGB4 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
High Mobility Group (HMG) proteins, a group of chromosomal proteins common to all eukaryotes, bind DNA in a non-sequence-specific fashion to promote chromatin function and gene regulation, helping with transcription, replication, recombination, and DNA repair. HMGB4 is HMG2-like, isoform 2, high-mobility group box 4. This gene can be found on Chromosome 1. HMGB4 contains two HMG-box regions, and is found in a variety of eukaryotic chromosomal proteins. HMGB proteins are phosphorylated to various extents. The existence of differentially modified forms increases the number of distinct HMGB protein variants in plant chromatin that may be adapted to certain functions.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HMGB4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge