Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NGB/neuroglobin Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1859R-A750
BOSSBS-1859R-A750EA 890 EUR InStock
BOSSBS-1859R-A750
Anti-NGB/neuroglobin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Involved in oxygen transport in the brain. Hexacoordinate globin, displaying competitive binding of oxygen or the distal His residue to the iron atom. Not capable of penetrating cell membranes. The deoxygenated form exhibits nitrite reductase activity inhibiting cellular respiration via NO-binding to cytochrome c oxidase. Involved in neuroprotection during oxidative stress. May exert its anti-apoptotic activity by acting to reset the trigger level of mitochondrial cytochrome c release necessary to commit the cells to apoptosis.

Type: Primary
Antigen: NGB/neuroglobin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-151/151
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge