Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RELMa/RELM alpha Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1884R-A680
BOSSBS-1884R-A680EA 874 EUR InStock
BOSSBS-1884R-A680
Anti-RELMa/RELM alpha Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
A family of resistin-like molecules (RELMs) has been identified in rodents and humans. RELM alpha belongs to a unique family of tissue-specific cytokines termed FIZZ (found in inflammatory zone) and RELM. The three known members of this family; Resistin, RELM alpha and RELM beta are 85-94 amino acid secreted proteins sharing a conserved C-terminal domain. RELM alpha and Resistin are secreted exclusively by adipocytes while RELM beta is expressed in the epithelium of the colon and small bowel. The RELMs together with resistin comprise a class of tissue-specific signaling molecules. The physiological role and molecular targets of RELM alpha are still unknown.

Type: Primary
Antigen: RELMa/RELM alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-111/111
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge