Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MMP8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1913R-A488
BOSSBS-1913R-A488EA 901 EUR InStock
BOSSBS-1913R-A488
Anti-MMP8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Matrix Metalloproteinase 8 (MMP8) is also known as neutrophil collagenase and collagenase 2. MMP8 degrades fibrillar collagens types I, II, III, aggrecan, serpins and alpha 2 macroglobulin. All collagenases cleave fibrillar collagens at one specific site resulting in generation of N terminal three quarter and C terminal one quarter fragments, which then denature to gelatin at body temperature. The substrate specificity of collagenases is variable: MMP1 degrades type III collagen more efficiently than type I or type II collagen, whereas MMP8 is more potent in degrading type I collagen than type III or type II collagen. MMP13, in turn degrades type II collagen 6 fold more efficiently than type I and type II collagens and displays almost 50 fold stronger gelatinolytic activity than MMP1 and MMP8. MMP8 is very similar to MMP1, sharing 57 % amino acid identity. Most cell types do not produce MMP8. Until recently, it was thought that MMP8 was produced exclusively by neutrophils, but it has also been detected in other cell types including arthritic chondrocytes and gingival fibroblasts. The human MMP8 gene has the chromosomal location of 11q22.2-22.3. MMP8 is heavily glycosylated, and the zymogen has a mass of 85 Kd. The zymogen is quickly activated to the 64 Kd form, and this breaks down to a cascade of active forms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MMP8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge