Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SCGB1A1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1935R-A647
BOSSBS-1935R-A647EA 893 EUR InStock
BOSSBS-1935R-A647
Anti-SCGB1A1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Uteroglobin belongs to the family of secretoglobins and is a secreted protein product of nonciliated bronchiolar Clara cells. There is convincing data suggesting it has phospholipase A2 inhibitory activity, as well as, a number of other immunomodulatory features including inhibition of interferon gamma signalling and Th1 vs. Th2 lymphocyte regulation. It was proposed as a potential peripheral marker of respiratory epithelial injury and bronchial dysfunction. Clara Cell Protein 16 concentrations have been determined in both serum and bronchoalveolar lavage fluid in numerous studies since 1994. In serum, its increase is associated with age, asbestos, nitrogen chloride and ozone exposure, sarcoidosis and high PEEP ventilation. Decreased serum CC16 levels are found after pulmonary resection, in silica-exposed workers, smokers and in asthma. Decreased CC16 concentrations were also found in the amniotic fluid of fetuses suffering from pulmonary hypoplasia caused by various mechanisms (diaphragmatic hernia, diabetic fetopathy, Turner and Down syndrome). In pleural effusions, the CC16 concentration appears to be associated with its diffusion from the lung as evidenced by high CC16 levels in cardiac pleural congestion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCGB1A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge