Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Zfp91 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1941R-A680
BOSSBS-1941R-A680EA 890 EUR InStock
BOSSBS-1941R-A680
Anti-Zfp91 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The protein encoded by ZFP91 is a member of the zinc finger family of proteins. This protein contains C2H2 type domains, which are the classical zinc finger domains found in numerous nucleic acid-binding proteins. May be involved in transcriptional regulation. May play an important role in cell proliferation and/or anti-apoptosis Found in all the examined tissues including brain, heart, kidney, lung, liver, spleen, thymus, skeletal muscle, ovary and testis. Belongs to the krueppel C2H2-type zinc-finger protein family.

Type: Primary
Antigen: Zfp91
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/570
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge