Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KLK9 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1967R-A647
BOSSBS-1967R-A647EA 893 EUR InStock
BOSSBS-1967R-A647
Anti-KLK9 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Kallikrein 9, also known as Kallikrein-Like 3 (KLK-L3), is a chymotrypsin-like serine proteinase. Kallikrein 9 was discovered as the locus for kallikreins on chromosome 19 was more fully mapped and found by similarity to the other tissue kallikreins. Kallikrein 9 has been found in the ovary, thymus, testis, prostate, skin, breast and neuronal tissues and is made by many cell lines in culture. Kallikrein 9 levels in breast cancer and uterine cancer patients have been reported to drop as the disease progresses, thus hK9 might be considered a favorable prognostic marker. Different splice variants of hK9 have been reported, although it is not yet known if they produce functional proteins. The full length Kallikrein 9 encodes for a 250 amino acid protein, with a predicted mass of 27.5 kDa and a pI of 7.53. The 234 amino acid form predicts a protein of 26 kDa with a pI of 9.76 and this quite basic pI might give the shorter form a very different function or localization. The shorter sequence also diverges before the catalytic serine residue, making it unlikely to be proteolytically active. Pre-pro-kallikrein 9 has the 17 amino acid signal sequence is removed before secretion, and the Pro-kallikrein 9 is activated to Kallikrein 9 by removal of the 5 amino acid propeptide domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLK9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge