Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LXN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1971R-FITC
BOSSBS-1971R-FITCEA 900 EUR InStock
BOSSBS-1971R-FITC
Anti-LXN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Latexin, a protein possessing inhibitory activity against rat carboxypeptidase A1 (CPA1) and CPA2, is expressed in a neuronal subset in the cerebral cortex and cells in other neural and non-neural tissues of rat. Although latexin also inhibits mast-cell CPA (MCCPA), the expression of latexin in rat mast cells has not previously been confirmed.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LXN
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge