Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PMP2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2032R-A350
BOSSBS-2032R-A350EA 893 EUR InStock
BOSSBS-2032R-A350
Anti-PMP2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
PMP2 is a small basic protein, belonging to the fatty-acid binding protein (FABP) family and is one of the major proteins of peripheral myelin. It appears to be involved in the transport of fatty acids or the metabolism of myelin lipids, and has been shown to have lipid-binding activity. PMP2 protein may have an important role in the organization of compact myelin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PMP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge