Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-E.coli DH-5 Alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2033R-A750
BOSSBS-2033R-A750EA 874 EUR InStock
BOSSBS-2033R-A750
Anti-E.coli DH-5 Alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
This antibody reacts with Escherichia coli HD5a (E.coli HD5a), Escherichia coli is a gram-negative bacillus that belongs to a larger group of Enterobacteriae - bacteria that inhabit the gastrointestinal tract. Although usually a harmless resident of the gut, some strains have the potential to cause serious problems, especially where there is an immature immune system or immunosuppression, or where the subtype of organism has acquired the ability to produce pathogenic toxins. E. coli DH5_ strain is an engineering strain widely used in biopharmaceuticals. According to the requirements of product quality inspection, it is necessary to detect the residual amount of DH5_ bacterial protein in biological products. This product is an anti-serum obtained by immunizing rabbits with DH5_ bacterial protein as an antigen, and purified by affinity chromatography to obtain antibody IgG, which can be used for ELISA, Western Blot and other experimental tests.

Type: Primary
Antigen: E.coli DH-5 Alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria, Others

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge