Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Methamphetamine Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2076R-CY5.5
BOSSBS-2076R-CY5.5EA 900 EUR InStock
BOSSBS-2076R-CY5.5
Anti-Methamphetamine Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Methamphetamine (METH) is closely related chemically to amphetamine (AMPH). METH is a potent central nervous system stimulant with additional peripheral sympathomimetic effects. METH and AMPH have been used clinically in the treatment of obesity, minimal brain dysfunction, narcolepsy, depression and to counter fatigue. They are also subjected to widespread abuse. METH is an indirect agonists. It causes the release of newly synthesized norepinephrine and dopamine and it blocks the re uptake of these transmitters from the synapse. This can lead to an increase in the concentration of catecholamines in the synapse as well as an overall increase in catecholaminergic activity in the brain. The mechanism of METH induced neurotoxicity for all monoaminergic cell types may lie primarily with the dopaminergic system in the striatum. It may also lie with the interaction between METH induced release of dopamine and its ability to inhibit monoamine oxidase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Methamphetamine
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge