Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RNF126 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2125R-CY5.5
BOSSBS-2125R-CY5.5EA 900 EUR InStock
BOSSBS-2125R-CY5.5
Anti-RNF126 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
E3 ubiquitin-protein ligase that regulates several biological processes through ubiquitination of various target proteins. Depending on the associated E2 ligase, mediates 'Lys-48'- and 'Lys-63'-linked polyubiquitination of substrates. Through their polyubiquitination, may play a role in the endosomal sorting and degradation of several membrane receptors including EGFR, FLT3, MET and CXCR4. May also be part of a BAG6-dependent quality control process ensuring that proteins of the secretory pathway that are mislocalized to the cytosol are degraded by the proteasome. May provide the ubiquitin ligase activity associated with the BAG6 complex and be responsible for ubiquitination of the mislocalized proteins and their targeting to the proteasome (PubMed:24981174). May also play a role in the endosomal recycling of IGF2R, the cation-independent mannose-6-phosphate receptor (PubMed:24275455). By ubiquitinating CDKN1A/p21 and targeting it for degradation, may also promote cell proliferation (PubMed:23026136). May monoubiquitinate AICDA (PubMed:23277564).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF126
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge