Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GPC4 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2159R-CY7
BOSSBS-2159R-CY7EA 900 EUR InStock
BOSSBS-2159R-CY7
Anti-GPC4 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Glypicans are glycosylphosphatidylinositol-anchored heparan sulphate proteoglycans, which are thought to modulate signalling by growth factors, including fibroblast growth factors and Wnts. Expression of glypican-4 has been reported in the mouse brain at embryonic day 7 and later stages.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Glypican 4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge