Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ADAR Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2168R-A555
BOSSBS-2168R-A555EA 893 EUR InStock
BOSSBS-2168R-A555
Anti-ADAR Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
ADAR1 converts adenosine to inosine in dsRNA, which destabilizes the dsRNA helix. This activity is important for various functions like site-specific RNA editing of transcripts of the glutamate receptors and modifying viral RNA genomes (which may be responsible for hypermutation of certain negative-stranded viruses, e.g., measles virus). ADAR1 also binds to short interfering RNAs (siRNA) without editing them and suppresses siRNA-mediated RNA interference. This protein is ubiquitously expressed, with the highest levels being found in brain and lung.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADAR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge