Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SMAD2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2224R-CY5.5
BOSSBS-2224R-CY5.5EA 905 EUR InStock
BOSSBS-2224R-CY5.5
Anti-SMAD2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
SMAD2 or Mothers against decapentaplegic homolog 2 is a polypeptide that, as its name describes, is a homolog of the Drosophila gene: "Mothers against decepentaplegic". It belongs to the SMAD family of proteins, which belong to the TGF-Beta superfamily of modulators. Like many other TGF-Beta family members SMAD2 is involved in cell signalling. SMAD2 modulates signals of activin and TGF-Beta's. It interacts with SMAD anchor for receptor activation (SARA). The binding of ligands causes the phosphorylation of the SMAD2 protein and the dissociation from SARA and the association with SMAD4. It is subsequently transferred to the nucleus where it forms complexes with other proteins and acts as a transcription factor. SMAD2 is a receptor regulated SMAD (R-SMAD) and is activated by bone morphogenetic protein type 1 receptor kinase. Smad2 (Mothers against decapentaplegic homolog 2; SMAD 2; Mothers against DPP homolog 2;)

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SMAD2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge