Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Cecropin Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2232R-CY7
BOSSBS-2232R-CY7EA 900 EUR InStock
BOSSBS-2232R-CY7
Anti-Cecropin Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Cecropins are produced by insects, particularly under conditions of infection. Cecropins are bioactive peptides that exhibit activities by interacting with membranes and forming transmembrane channels that allow the free flow of electrolytes, metabolites and water across the phospholipid bilayers. Cecropins A, B and D are close homologues consisting of 35-39 residues. They are found in the pupae of the cecropin moth, but related homologues named lepodopteran, bactericidin, moricin and sarcotoxin are produced by other insects.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Cecropin
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge