Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DHRS3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2286R-A750
BOSSBS-2286R-A750EA 890 EUR InStock
BOSSBS-2286R-A750
Anti-DHRS3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
DHRS3 belongs to the short-chain dehydrogenases/reductases (SDR) family. It catalyses the reduction of all-trans-retinal to all-trans-retinol in the presence of NADPH. DHRS3 is located in a region of chromosome 1 which is often deleted in aggressive neuroblastoma tumors. There are two named isoforms.

Type: Primary
Antigen: DHRS3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/302
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge