Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HSD3B7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2366R-A750
BOSSBS-2366R-A750EA 890 EUR InStock
BOSSBS-2366R-A750
Anti-HSD3B7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The 3-beta-HSD enzymatic system plays a crucial role in the biosynthesis of all classes of hormonal steroids. HSD VII is active against four 7-alpha-hydroxylated sterols. Does not metabolize several different C(19/21) steroids as substrates. Involved in bile acid synthesis.

Type: Primary
Antigen: HSD3B7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 281-369/369
Host: Rabbit
Isotype: IgG
Reactivity: Rat; Others

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge