Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GPER1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2367R-CY7
BOSSBS-2367R-CY7EA 893 EUR InStock
BOSSBS-2367R-CY7
Anti-GPER1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
A member of the family of glycosphingolipids, monosialoanglioside acts as a receptor and antigen for cholera. GM1 ganglioside, one of the glycosphingolipids widely distributed in all tissues, occurs in highest concentrations in the central nervous system (CNS). It is primarily located in the outer surface of the mammalian cell's plasma membrane and in synaptic membranes of the CNS. GM1 ganglioside modulates a number of cell surface and receptor activities as well as neuronal differentiation and development, protein phosphorilation and synaptic function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ganglioside GM1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge