Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-C9ORF172 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2374R-PE
BOSSBS-2374R-PEEA 919 EUR InStock
BOSSBS-2374R-PE
Anti-C9ORF172 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
TAP is an integral transmembrane protein involved in the transport of antigens from the cytoplasm to the endoplasmic reticulum for association with MHC class I molecules. It is a heterodimer of TAP1 and TAP2, and the peptide-binding site is shared between the cytoplasmic loops of TAP1 and TAP2. TAP is inducible by interferon gamma and belongs to the ABC transporter family, MDR subfamily. TAP also acts as a molecular scaffold for the final stage of MHC class I folding, namely the binding of peptide. Nascent MHC class I molecules associate with TAP via tapasin. TAP is inhibited by the covalent attachment of herpes simplex virus ICP47 protein, which blocks the peptide-binding site of TAP. It is inhibited by human cytomegalovirus US6 glycoprotein, which binds to the lumenal side of the TAP complex and inhibits peptide translocation by specifically blocking ATP-binding to TAP and prevents the conformational rearrangement of TAP induced by peptide binding.

Recommended Dilutions: Flow Cytometry: 1:20-100

Type: Primary
Antigen: C9ORF172
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge