Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-APLN Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2425R-CY5
BOSSBS-2425R-CY5EA 900 EUR InStock
BOSSBS-2425R-CY5
Anti-APLN Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Apelin is a neuropeptide expressed in the supraoptic and paraventricular nuclei and is an endogenous ligand for APJ, a G protein-coupled orphan receptor which is an alternative coreceptor with CD4 for HIV-1. Apelin and APJ are ubiquitously expressed in peripheral tissues, with highest levels reported for heart and lungs, as well several regions within the central nervous system. The actions of apelin remain largely unknown: Apelin inhibits HIV-1 entry in cells coexpressing CD4 and APJ; the oral intake of Apelin in colostrum and breast milk could have a role in the modulation of the immune responses in neonates; more recent studies have also indicated a role for Apelin in the central control of body fluid homeostasis, by influencing AVP release and drinking behavior. In the cardiovascular system several actions of Apelin have been described, including endothelium-dependent vasodilatation, vasoconstriction through direct action on smooth muscle and positive inotropism.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APLN
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge