Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TWIST1 and TWIST2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2441R-A680
BOSSBS-2441R-A680EA 899 EUR InStock
BOSSBS-2441R-A680
Anti-TWIST1 and TWIST2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
TWIST1 and TWIST2 are basic helix-loop-helix transcriptional repressors that bind to E boxes in gene promoters, acting as master regulators in a variety of biological processes, including organogenesis, osteogenesis, cancer progression and hematopoietic cell development. Both TWIST1 and TWIST2 are found to act as Homodimers or Heterodimers with another bHLH protein. TWIST1 Acts as a transcriptional regulator. Inhibits myogenesis by sequestrating E proteins, inhibiting trans-activation by MEF2, and inhibiting DNA-binding by MYOD1 through physical interaction. TWIST1 also represses expression of proinflammatory cytokines such as TNFA and IL1B. TWIST2 inhibits transcriptional activation by MYOD1, MYOG, MEF2A, and MEF2C. Twist2 inhibits the premature or ectopic differentiation of preosteoblast cells during osteogenesis.

Type: Primary
Antigen: TWIST1 and TWIST2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 71-170/202
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge