Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CCL9 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2477R-A350
BOSSBS-2477R-A350EA 901 EUR InStock
BOSSBS-2477R-A350
Anti-CCL9 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
This chemotactic factor that attracts monocytes, lymphocytes, basophils and eosinophils may play a role in neoplasia and inflammatory host responses and can bind heparin. Its highest expression is found in the small intestine and peripheral blood cells. It is induced by interferon gamma, mitogens and interleukin 1 and belongs to the intercrine beta (chemokine CC) family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCL9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge