Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-JAK2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2485R-CY5.5
BOSSBS-2485R-CY5.5EA 905 EUR InStock
BOSSBS-2485R-CY5.5
Anti-JAK2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
JAK2 (Janus Activating Kinase 2) is a tyrosine kinase of the non-receptor type, that associates with the intracellular domains of cytokine receptors; JAK2 is the predominant JAK kinase activated in response to several growth factors and cytokines such as IL-3, GM-CSF and erythropoietin; it has been found to be constitutively associated with the prolactin receptor and is required for responses to gamma interferon. Ligand binding to a variety of cell surface receptors (e.g., cytokine, growth factor, GPCRs) leads to an association of those receptors with JAK proteins, which are then activated via phosphorylation on tyrosines 1007 and 1008 in the kinase activation loop. Activated JAK proteins phosphorylate and activate STAT (signal transducers and activators of transcription) proteins, which then dimerize and translocate to the nucleus. Once in the nucleus, STAT proteins bind to DNA and modify the transcription of various genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: JAK2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge