Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CLEC6A Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2566R-A488
BOSSBS-2566R-A488EA 893 EUR InStock
BOSSBS-2566R-A488
Anti-CLEC6A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Dectin 2 is a type II transmembrane protein that is a member of the C type lectin superfamily. In mouse, dectin 2 is predominantly expressed on tissue macrophages and some dendritic cells. Significant expression of Dectin 2 has been reported on macrophages in the red pulp and marginal zones of the spleen, kupffer cells in the liver and alveolar macrophages in the lung. Peripheral blood monocytes express low levels of dectin 2 but transient up regulation of expression has been demonstrated on monocytes at sites of inflammation. The function of dectin 2 has not been fully determined but studies suggest a possible role in immune surveillance.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLEC6A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge