Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Lymphocyte Activation Gene 3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2646R-A680
BOSSBS-2646R-A680EA 874 EUR InStock
BOSSBS-2646R-A680
Anti-Lymphocyte Activation Gene 3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Lymphocyte-activation protein 3 (LAG-3), also known as CD223, belongs to the immunoglobulin superfamily and contains 4 extracellular Ig-like domains. LAG-3 binds to HLA class-II antigens and is involved in lymphocyte activation. It is found on the cell surface of activated NK and T-lymphocytes. The sequence data, exon/intron organisation, and chromosomal localization all indicate a close relationship of LAG-3 to CD4.

Type: Primary
Antigen: Lymphocyte Activation Gene 3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/525
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge