Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2661R-A750
BOSSBS-2661R-A750EA 890 EUR InStock
BOSSBS-2661R-A750
Anti-MD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
MD1 is a 28 kDa molecule that is associated at the cell surface with RP105 (CD180). RP105 is a leucine-rich repeat (LRR) molecule that is expressed on B lymphocytes. It was first identified by a mAb that protects spleen B cells from irradiation-induced apoptosis. LRR proteins, such as Toll receptors, have a role in innate immunity. MD1 is primarily expressed by B cells, dendritic cells and monocytes, and promotes the recognition of, and subsequent signaling by LPS in the innate immune system. MD1 does not have any homology to other molecules. MD1 when expressed alone behaves as a secretory protein. Expression of MD1 in a cell increases the expression of RP105.

Type: Primary
Antigen: MD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 65-162/162
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge