Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PTPRJ Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2675R-A488
BOSSBS-2675R-A488EA 893 EUR InStock
BOSSBS-2675R-A488
Anti-PTPRJ Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
a-Fodrin is a universally expressed membrane associated cytoskeletal protein consisting of alpha and beta subunits. The protein is important for maintaining normal membrane structure and supporting cell surface protein function. Alpha Fodrin is one of the important targets cleaved by caspases during apoptosis. The full length 240 kDa protein can be cleaved at several sites within its sequence by activated caspases generating N terminal 150 kDa, C terminal 120 kDa, and 35 kDa major products. Cleavage of alpha Fodrin leads to membrane malfunction and cell shrinkage.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTPRJ
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge