Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CRLF2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2727R-A350
BOSSBS-2727R-A350EA 893 EUR InStock
BOSSBS-2727R-A350
Anti-CRLF2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
TSLPR is a type I membrane receptor that forms a functional heterodimeric complex with IL7R to bind TSLP. The TSLP R contains a WSXWS motif required for proper protein folding and a box1 motif important for association with the JAKs. TSLPR has a predicted molecular weight approximately 41 kD, and two further isoforms have been reported that are produced by alternative splicing. The TSLPR is expressed preferentially in myeloid cells including dendritic cells and activated monocytes, and is weakly expressed in T cells. Expression has also been reported in heart, skeletal muscle, and kidney tissues. TSLP binding to the heterodimeric functional receptor (TSLPR and IL7R) activates JAK2, STAT3 and STAT5 to stimulate cell proliferation. Ligand receptor interactions haves been implicated in the development of the hematopoietic system, dendritic cell maturation, and the maintenance and polarization of human Th2 memory T cells in allergic diseases.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRLF2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge