Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CRK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2763R-FITC
BOSSBS-2763R-FITCEA 900 EUR InStock
BOSSBS-2763R-FITC
Anti-CRK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Crk p38 (CrkII) is an isoform of the protein Crk. CrkI and CrkII are produced from the same crk gene by alternative splicing. The two isoforms differ in their biological activities with CrkII having less transforming activity than CrkI.Crk p38 is a member of an adapter protein family that binds to several tyrosine-phosphorylated proteins. It has several SH2 and SH3 domains (src-homology domains) and is involved in several signaling pathways, recruiting cytoplasmic proteins in the vicinity of tyrosine kinase through SH2-phosphotyrosine interaction. The N-terminal SH2 domain of this protein functions as a positive regulator of transformation whereas the C-terminal SH3 domain functions as a negative regulator of transformation. Crk is believed to be a regulator of invasive responses because increased levels of the protein have been observed in multiple human cancers. In vivo studies have demonstrated that decreased levels of Crk remarkably inhibits tumor formation and its invasive growth.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRK
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge