Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EDG6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2883R-A750
BOSSBS-2883R-A750EA 890 EUR InStock
BOSSBS-2883R-A750
Anti-EDG6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
This protein belongs to a G protein coupled heptahelical receptor subfamily named Endothelial Cell Differentiation Genes (EDG)that act as receptors for biologically active lysophospholipids. This group consists of two receptor subgroups specific for S1P and LPA respectively. EDG6 is the receptor for lysophospholipid sphingosine 1 phosphate (S1P). S1P elicits diverse physiological effect on most types of cells and tissues. EDG6 may be involved in cell migration processes that are specific for lymphocytes.

Type: Primary
Antigen: EDG6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 301-384/384
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge